Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals
Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number 039448.03
also known as 039448-03
Price (USD)/ Each
91.65
Online exclusive
102.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
91.65
Online exclusive
102.00 
Save 10.35 (10%)
Add to cart
Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals
Catalog number039448.03
Price (USD)/ Each
91.65
Online exclusive
102.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS42196-31-6
IUPAC Namepalladium(2+) ditrifluoroacetate
Molecular FormulaC4F6O4Pd
InChI KeyPBDBXAQKXCXZCJ-UHFFFAOYSA-L
SMILES[Pd++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥96.0%
Appearance (Color)Brown
Infrared spectrumConforms
FormPowder or crystals or crystalline powder
Palladium(II) trifluoroacetate is used in the preparation of allyl-palladium complexes. It acts as a catalyst used in the mild decarboxylation of electron-rich aromatic acids and the direct cross-coupling of unactivated arenes. Further, it is involved in the selective allylic oxidation of geranylacetone and other olefins to their allyl acetates in order to prepare keto alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only