Thermo Scientific Chemicals

Nickel naphthenate, Ni 5-12%, Thermo Scientific Chemicals

Catalog number: 039455.A1
1 kg, Each
Thermo Scientific Chemicals

Nickel naphthenate, Ni 5-12%, Thermo Scientific Chemicals

Catalog number: 039455.A1
1 kg, Each
Quantity
Catalog number: 039455.A1
also known as 039455-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61788-71-4
IUPAC Name
nickel(2+) bis(naphthalene-2-carboxylate)
Molecular Formula
C22H14NiO4
InChI Key
UIEKYBOPAVTZKW-UHFFFAOYSA-L
SMILES
[Ni++].[O-]C(=O)C1=CC2=CC=CC=C2C=C1.[O-]C(=O)C1=CC2=CC=CC=C2C=C1
Appearance (Color)
Green
Form
Viscous liquid
Assay (unspecified)
Wt % Ni: 5 to 12%

Description

Applied as a rubber catalyst. The product is a aid siccative and also acts as a stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text