Thermo Scientific Chemicals

Barium tetracyanoplatinate(II) tetrahydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 039463.03
1 g, Each
Thermo Scientific Chemicals

Barium tetracyanoplatinate(II) tetrahydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 039463.03
1 g, Each
Quantity
Catalog number: 039463.03
also known as 039463-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13755-32-3
IUPAC Name
barium(2+) tetracyanoplatinumdiuide tetrahydrate
Molecular Formula
C4H12BaN4O4Pt
InChI Key
QNTHMRLOHDQAOV-UHFFFAOYSA-N
SMILES
O.O.O.O.[Ba++].[Pt--].C#N.C#N.C#N.C#N
Assay (special)
Platinum (Pt): 37.78-38.74%
Appearance (Color)
Yellow
Form
Powder
Assay from Supplier's CofA
≥99.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text