Niobium silicide, 99.5% (metals basis), Thermo Scientific Chemicals
Niobium silicide, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium silicide, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 039471.14
also known as 039471-14
Price (USD)/ Each
81.60
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
81.60
Add to cart
Niobium silicide, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number039471.14
Price (USD)/ Each
81.60
-
Add to cart
Chemical Identifiers
CAS12060-34-3
IUPAC Namepentaniobium trisilicon
Molecular FormulaNb5Si3
InChI KeyYSJRXBWBYGDQAI-UHFFFAOYSA-N
SMILES[Si].[Si].[Si].[Nb].[Nb].[Nb].[Nb].[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
The Nb-based RMIC’s have much higher potential application temperatures, because of their dramatically higher melting temperatures. It is used in high temperature in situ composites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only