Thermo Scientific Chemicals

Niobium silicide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 039471.22
100 g, Each
Thermo Scientific Chemicals

Niobium silicide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 039471.22
100 g, Each
Quantity
Catalog number: 039471.22
also known as 039471-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12060-34-3
IUPAC Name
pentaniobium trisilicon
Molecular Formula
Nb5Si3
InChI Key
YSJRXBWBYGDQAI-UHFFFAOYSA-N
SMILES
[Si].[Si].[Si].[Nb].[Nb].[Nb].[Nb].[Nb]
Total Metal Impurities
0.5% max.

Description

The Nb-based RMIC’s have much higher potential application temperatures, because of their dramatically higher melting temperatures. It is used in high temperature in situ composites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text