Nickel(II) octanoate, in mineral spirits (8% Ni), Thermo Scientific Chemicals
Nickel(II) octanoate, in mineral spirits (8% Ni), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) octanoate, in mineral spirits (8% Ni), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 039479.22
also known as 039479-22
Price (USD)/ Each
175.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
175.00
Add to cart
Nickel(II) octanoate, in mineral spirits (8% Ni), Thermo Scientific Chemicals
Catalog number039479.22
Price (USD)/ Each
175.00
-
Add to cart
Specifications
Chemical Name or MaterialNickel(II) octanoate
Name NoteIn mineral spirits
CAS4995-91-9
Health Hazard 1H226-H304-H317-H334-H340-H350i-H360D-H372
Health Hazard 2GHS H Statement
H334-H340-H350-H360-H372-H304-H226-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
May cause an allergic skin reaction.
View more
Nickel(II) octanoate is used to prepare nickel based catalysts using niobia modified silica as support. It is also used as a catalyst and involved in the treatment of qaiyarah heavy crude oil, petrolene using ethanol as a solvent and reductant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only