Nickel(II) cyanide tetrahydrate, Thermo Scientific Chemicals
Nickel(II) cyanide tetrahydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) cyanide tetrahydrate, Thermo Scientific Chemicals

For Metallurgy, Nickel-Plating
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 039480.09
also known as 039480-09
Price (USD)/ Each
191.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
191.00
Add to cart
Nickel(II) cyanide tetrahydrate, Thermo Scientific Chemicals
Catalog number039480.09
Price (USD)/ Each
191.00
-
Add to cart
Chemical Identifiers
CAS13477-95-7
IUPAC Namenickel(2+) bis(iminomethanide) tetrahydrate
Molecular FormulaC2H8N2NiO4
InChI KeyVHHVAACTKRUWEK-UHFFFAOYSA-N
SMILESO.O.O.O.[Ni++].[C-]#N.[C-]#N
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content1% max
Applied in metallurgy, nickel-plating industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only