Thermo Scientific Chemicals

Zirconium titanium oxide, 99.5% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals

Catalog number: 039490.22
100 g, Each
Thermo Scientific Chemicals

Zirconium titanium oxide, 99.5% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals

Catalog number: 039490.22
100 g, Each
Quantity
Catalog number: 039490.22
also known as 039490-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12036-70-3
Impurity content
Hf <75ppm
Total Metal Impurities excluding Hf
0.5% max.

Description

Suitable for glass, optic and ceramic applications, advanced electronics and in light weight structural components in aerospace and electrochemical applications such as fuel cells in which they exhibit ionic conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text