Iridium(IV) bromide, Premion™, 99.99% (metals basis), Ir 37.1% min, Thermo Scientific Chemicals
Iridium(IV) bromide, Premion™, 99.99% (metals basis), Ir 37.1% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iridium(IV) bromide, Premion™, 99.99% (metals basis), Ir 37.1% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
0.5 g
Catalog number 039493.04
also known as 039493-04
Price (USD)/ Each
-
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS7789-64-2
Molecular FormulaBr4Ir
InChI KeyIDTZIDJMCPQPPV-UHFFFAOYSA-N
SMILES[Br].[Br].[Br].[Br].[Ir]
Molecular Weight (g/mol)511.83
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only