Thermo Scientific Chemicals

Osmium(IV) oxide, Os 83% min, Thermo Scientific Chemicals

Catalog number: 039497.01
0.25 g, Each
Thermo Scientific Chemicals

Osmium(IV) oxide, Os 83% min, Thermo Scientific Chemicals

Catalog number: 039497.01
0.25 g, Each
Quantity
Catalog number: 039497.01
also known as 039497-01
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12036-02-1
IUPAC Name
dioxoosmium
Molecular Formula
O2Os
InChI Key
XSXHWVKGUXMUQE-UHFFFAOYSA-N
SMILES
O=[Os]=O
Form
Powder
Assay (unspecified)
≥83.0% Osmium
Appearance (Color)
Black to black-brown to grey

Description

The osmium oxide/hexacyanoruthenate films modified electrodes plays an important role for the electrocatalytic reaction of catecholamines and sulfur oxoanions. Used as catalyst. Metallic osmium or an osmium oxide-ruthenium cyanide film modified graphite electrodes plays an essential role for the determination of biogenic amines from electrocatalytic responses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text