Thermo Scientific Chemicals

Potassium orthovanadate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 039502.14
25 g, Each
Thermo Scientific Chemicals

Potassium orthovanadate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 039502.14
25 g, Each
Quantity
Catalog number: 039502.14
also known as 039502-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14293-78-8
IUPAC Name
potassium vanadium trioxidandiide
Molecular Formula
KO3V
InChI Key
WAMGQABPPBZHJS-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[K+].[V]
Total Metal Impurities
0.1% max.

Description

It is employed as in chemical fertilizer industries and used as corrosion inhibitor. It is also used for ammonia synthesis. Used as a catalyst in oil & gas industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text