Samarium(III) hydroxide hydrate, 99% (REO), Thermo Scientific Chemicals
Samarium(III) hydroxide hydrate, 99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Samarium(III) hydroxide hydrate, 99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 039512.06
also known as 039512-06
Price (USD)/ Each
52.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
52.60
Add to cart
Samarium(III) hydroxide hydrate, 99% (REO), Thermo Scientific Chemicals
Catalog number039512.06
Price (USD)/ Each
52.60
-
Add to cart
Chemical Identifiers
CAS20403-06-9
IUPAC Namesamarium(3+) trihydroxide
Molecular FormulaH3O3Sm
InChI KeyBCYBEIXXOVNETJ-UHFFFAOYSA-K
SMILES[OH-].[OH-].[OH-].[Sm+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities1% max.
Samarium hydroxide (Sm(OH)3) is an important samarium compound with many potential applications in electronics, optics and catalysis. It is used as a chemical intermediate. It is used in the separation and preconcentration procedures for the determination of lead using spectrometric techniques. It has specialized uses in glass, phosphors, lasers, and thermoelectric devices and it is mainly applied in Neutron absorption.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only