Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For fast, reliable anion eluent preparation, the ready-to-dilute Thermo Scientific™ Dionex™ AS4A Eluent Concentrate; Sodium Carbonate/Bicarbonate Concentrate will save you time and eliminate the worry of potential contamination or preparation error. For use with Dionex™ IonPac™ AS4A, AS4A-SC, and AS9-SC columns, simply prepare eluent by dissolving the concentrate in deionized water. The 100x concentrate is made with only high purity, reagent grade chemicals and water (18 MO-cm) and quality checked for ionic contamination. The concentrate contains 0.18M carbonate and0.17M bicarbonate.
For Use With (Equipment) | IonPac AS4 A, AS4 A-SC and AS9-SC Columns |
Type | Eluent Concentrate |
Standard Concentration | 0.18, 0.17 M |
Concentration | 100 X |
Hazards | Irritant |
Standard Components | Sodium Carbonate, Sodium Bicarbonate Aqueous Solution |
Quantity | 500 mL |
Description | AS4 A Sodium Carbonate/Bicarbonate Concentrate; 100 X; 500 mL |
Storage | Room Temperature |
Unit Size | Each 1 |
Catalog Number | Specifications | Unit Size | For Use With (Equipment) | Quantity | Concentration | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
039513 | Each 1 | IonPac AS4 A, AS4 A-SC and AS9-SC Columns | 500 mL | 100 X | 183.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|