Thermo Scientific Chemicals

Triphenylphosphine-3,3',3″-trisulfonic acid trisodium salt hydrate, tech. 85%, Thermo Scientific Chemicals

Catalog number: 039538.01
0.25 g, Each
Thermo Scientific Chemicals

Triphenylphosphine-3,3',3″-trisulfonic acid trisodium salt hydrate, tech. 85%, Thermo Scientific Chemicals

Catalog number: 039538.01
0.25 g, Each
Quantity
Catalog number: 039538.01
also known as 039538-01
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
335421-90-4
IUPAC Name
trisodium 3-[bis(3-sulfonatophenyl)phosphanyl]benzene-1-sulfonate
Molecular Formula
C18H12Na3O9PS3
InChI Key
MYAJTCUQMQREFZ-UHFFFAOYSA-K
SMILES
[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC(=CC=C1)P(C1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=CC(=C1)S([O-])(=O)=O
Form
Powder
Appearance (Color)
White
Assay by NMR
> 85.0%

Description

Triphenylphosphine-3,3',3″-trisulfonic acid trisodium salt hydrate is useful for the preparation of water-soluble organometal catalysts, which finds application in the hydroformylation of propene. It acts as a Lewis base, which is stronger than triphenylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text