Zirconium(IV) tert-butoxide, 99.99% (metals basis), Thermo Scientific Chemicals
Zirconium(IV) tert-butoxide, 99.99% (metals basis), Thermo Scientific Chemicals
Zirconium(IV) tert-butoxide, 99.99% (metals basis), Thermo Scientific Chemicals
Zirconium(IV) tert-butoxide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) tert-butoxide, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039546.09
also known as 039546-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
Beilstein3681870
CAS2081-12-1
Chemical Name or MaterialZirconium(IV) tert-butoxide
ColorColorless
View more
It finds it application as a reagent for the preparation of heterometallic tin(II) or lead(II) zirconium alkoxides. It is used as a catalyst in cyanation process. Zirconium tert-butoxide (ZTB) precursor is used to deposit thin films of Zirconia and other zirconium containing films by atomic layer deposition and chemical vapor deposition methods. Zirconium oxide thin films can also be grown at low temperatures, ranging from 150°C to 300°C, by the presence of moisture along with ZTB or by UV-enhanced atomic layer deposition (UV-ALD) process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link