Pentamethylcyclopentadienylzirconium trichloride, 99%, Thermo Scientific Chemicals
Pentamethylcyclopentadienylzirconium trichloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pentamethylcyclopentadienylzirconium trichloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039551.03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS75181-07-6
Molecular FormulaC10H15Cl3Zr
InChI KeyXZRIJZMMTDVZIR-UHFFFAOYSA-K
SMILESCl[Zr](Cl)Cl.Cc1c(C)c(C)[c-](C)c1C
Molecular Weight (g/mol)332.80
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
FormPowder
Assay (Titration ex Chloride)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only