Thermo Scientific Chemicals

Bis(trimethylsilyl)sulfide, 98%, Thermo Scientific Chemicals

Catalog number: 039556.06
5 g, Each
Thermo Scientific Chemicals

Bis(trimethylsilyl)sulfide, 98%, Thermo Scientific Chemicals

Catalog number: 039556.06
5 g, Each
Quantity
Catalog number: 039556.06
also known as 039556-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3385-94-2
IUPAC Name
trimethyl[(trimethylsilyl)sulfanyl]silane
Molecular Formula
C6H18SSi2
InChI Key
RLECCBFNWDXKPK-UHFFFAOYSA-N
SMILES
C[Si](C)(C)S[Si](C)(C)C
Appearance (Color)
Clear, colorless
Transition Temperature
Liquid
Assay (GC)
≥97.5%

Description

Bis(trimethylsilyl)sulfide is used as a silylating agent in the synthesis of pseudohalides, trifluoroacetate and tetramethylsilyl halides. It is used in the transformation of oxides and chlorides into corresponding sulfides. It is also used in the preparation of dimethyltrisulfane as well as thiones from aldehydes and ketones. Further, it is used as a reducing agent to reduce aromatic nitro compounds to amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text