Specifications
Chemical Name or MaterialZinc hexafluoro-2,4-pentanedionate dihydrate
Melting Point157°C to 158°C
CAS16743-33-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more