Thermo Scientific Chemicals

Zinc 2-ethylhexanoate, ca 80% in mineral spirits (17-19% Zn), Thermo Scientific Chemicals

Catalog number: 039561.0E
2500 g, Each
Thermo Scientific Chemicals

Zinc 2-ethylhexanoate, ca 80% in mineral spirits (17-19% Zn), Thermo Scientific Chemicals

Catalog number: 039561.0E
2500 g, Each
Quantity
Catalog number: 039561.0E
also known as 039561-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
136-53-8
IUPAC Name
zinc(2+) bis(2-ethylhexanoate)
Molecular Formula
C16H30O4Zn
InChI Key
IFNXAMCERSVZCV-UHFFFAOYNA-L
SMILES
[Zn++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Specific Gravity
1.050-1.090 @ 25?C
Elemental Analysis
Zn: 17.80-18.20%
Color (Gardner Scale)
≤6
Viscosity
A5 - I (Gardner Holdt @ 25?C)
Residue on Evaporation
75.00-85.00%

Description

It is applied as a catalyst for condensation-cure silicones and as a vulcanization activator in industries. It is also employed in PVC heat stabilizer, polyurethane catalyst and lubricant additive. They are used as catalysts in paints and coatings, in polyurethanes, greases, and as wetting agents for pigments. Also used in the formulation of lubricants and greases, as hardener for epoxy resins, to make rubber and plastic products, and for refining crude oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text