Yttrium(III) sulfide, 99.9% (REO), Thermo Scientific Chemicals
Yttrium(III) sulfide, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium(III) sulfide, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 039565.03
also known as 039565-03
Price (USD)/ Each
72.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
72.80
Add to cart
Yttrium(III) sulfide, 99.9% (REO), Thermo Scientific Chemicals
Catalog number039565.03
Price (USD)/ Each
72.80
-
Add to cart
Chemical Identifiers
CAS12039-19-9
IUPAC Namediyttrium(3+) trisulfanediide
Molecular FormulaS3Y2
InChI KeyBPMRLDMEAPSVQN-UHFFFAOYSA-N
SMILES[S--].[S--].[S--].[Y+3].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only