Thermo Scientific Chemicals

Bis(pentamethylcyclopentadienyl)titanium dichloride, Thermo Scientific Chemicals

Catalog number: 039570.03
1 g, Each
Thermo Scientific Chemicals

Bis(pentamethylcyclopentadienyl)titanium dichloride, Thermo Scientific Chemicals

Catalog number: 039570.03
1 g, Each
Quantity
Catalog number: 039570.03
also known as 039570-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
11136-36-0
IUPAC Name
titanium(4+) bis(pentamethylcyclopenta-1,3-dien-1-ide) dichloride
Molecular Formula
C20H30Cl2Ti
InChI Key
ZBFBXTFQCKIUHU-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Ti+4].CC1=[C-]C(C)(C)C(C)=C1C.CC1=[C-]C(C)(C)C(C)=C1C
Assay from Supplier's CofA
≥98.0%
Appearance (Color)
Red to red-brown to brown
Form
Powder or crystals or crystalline powder

Description

It is used as an intermediate in the synthesis of troticenyl monophosphanes and [2]troticenophanes with C-P and C-Si bridges.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text