Strontium oxalate, 95%, Thermo Scientific Chemicals
Strontium oxalate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium oxalate, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 g
Catalog number 039573.36
also known as 039573-36
Price (USD)/ Each
145.65
Online exclusive
162.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
145.65
Online exclusive
162.00 
Save 16.35 (10%)
Add to cart
Strontium oxalate, 95%, Thermo Scientific Chemicals
Catalog number039573.36
Price (USD)/ Each
145.65
Online exclusive
162.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS814-95-9
IUPAC Namestrontium(2+) oxalate
Molecular FormulaC2O4Sr
InChI KeyKQAGKTURZUKUCH-UHFFFAOYSA-L
SMILES[Sr++].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (unspecified)> 94.0%
FormPowder or lump
Loss on Drying0.25% Max
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only