Ytterbium(III) isopropoxide, Thermo Scientific Chemicals
Ytterbium(III) isopropoxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ytterbium(III) isopropoxide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 039579.06
also known as 039579-06
Price (USD)/ Each
354.65
Online exclusive
394.00 
Save 39.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
354.65
Online exclusive
394.00 
Save 39.35 (10%)
Add to cart
Ytterbium(III) isopropoxide, Thermo Scientific Chemicals
Catalog number039579.06
Price (USD)/ Each
354.65
Online exclusive
394.00 
Save 39.35 (10%)
-
Add to cart
Chemical Identifiers
CAS6742-69-4
IUPAC Nametris(propan-2-ol) ytterbium
Molecular FormulaC9H24O3Yb
InChI KeyIVGMSSQGEVYWID-UHFFFAOYSA-N
SMILES[Yb].CC(C)O.CC(C)O.CC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale cream
Total Metal Impurities≤1.0%
Yttrium(III) tris(isopropoxide) is a catalyst for stereoselective conjugate addition reactions, ring-opening polymerization, generation of reactive enolates, allylic substitution and cyano-phosphorylation of enones and synthesis of nanocomposites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only