Thermo Scientific Chemicals

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)ytterbium(III), 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 039582.06
5 g, Each
Thermo Scientific Chemicals

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)ytterbium(III), 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 039582.06
5 g, Each
Quantity
Catalog number: 039582.06
also known as 039582-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18323-96-1
IUPAC Name
ytterbium(3+) tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-dioxooctan-4-ide)
Molecular Formula
C30H30F21O6Yb
InChI Key
QYXREIJIWYJRCU-UHFFFAOYSA-N
SMILES
[Yb+3].CC(C)(C)C(=O)[CH-]C(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(=O)[CH-]C(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(=O)[CH-]C(=O)C(F)(F)C(F)(F)C(F)(F)F
Form
Crystalline powder
Appearance (Color)
Pale yellow
Chloride content
< 0.1%
Assay (unspecified)
> 98.5%

Description

It is a reactant involved in the synthesis of rare earth metal complexes for separation of lanthanide metals, conformational mobility of substituted 2-methoxychalcones under the action of lanthanide shift reagents, NMR titration on cation-π complexes of bowl-shaped polycyclic aromatic hydrocarbons, specific recognition of chiral amino alcohols. It finds its application as catalysis and synthesis, optics & glasses, healthcare & biochemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text