Thermo Scientific Chemicals

Vanadium(IV) etioporphyrin III oxide, Thermo Scientific Chemicals

Catalog number: 039586.MF
50 mg, Each
Thermo Scientific Chemicals

Vanadium(IV) etioporphyrin III oxide, Thermo Scientific Chemicals

Catalog number: 039586.MF
50 mg, Each
Quantity
Catalog number: 039586.MF
also known as 039586-MF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15709-03-2
UV/Vis
Conforms ( shows three maxima at ~570, 532 and 406 nm)
Appearance (Color)
Purple
Form
solid

Description

Vanadium(IV) etioporphyrin III oxide is used to check the trapping efficiency of the external AlOxlayer in AlOxcoating of ultrastable Zeolite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text