Thermo Scientific Chemicals

Vanadium(IV) oxide phthalocyanine, Thermo Scientific Chemicals

Catalog number: 039587.04
2 g, Each
Thermo Scientific Chemicals

Vanadium(IV) oxide phthalocyanine, Thermo Scientific Chemicals

Catalog number: 039587.04
2 g, Each
Quantity
Catalog number: 039587.04
also known as 039587-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13930-88-6
IUPAC Name
2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide; oxovanadiumbis(ylium)
Molecular Formula
C32H16N8OV
InChI Key
YRZZLAGRKZIJJI-UHFFFAOYSA-N
SMILES
O=[V++].[N-]1C2=NC3=C4C=CC=CC4=C([N-]3)N=C3N=C(N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
Appearance (Color)
Dark blue to purple
Form
Powder
Assay from Suppliers CofA
≥80.0%
Infrared spectrum
Conforms

Description

It is used as an organic light-emitting diode intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text