Titanium(IV) oxide sulfate sulfuric acid hydrate, Thermo Scientific Chemicals
Titanium(IV) oxide sulfate sulfuric acid hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium(IV) oxide sulfate sulfuric acid hydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039591.A3
also known as 039591-A3
Price (USD)/ Each
301.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
355.00 
Save 53.35 (15%)
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
301.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
355.00 
Save 53.35 (15%)
Add to cart
Titanium(IV) oxide sulfate sulfuric acid hydrate, Thermo Scientific Chemicals
Catalog number039591.A3
Price (USD)/ Each
301.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
355.00 
Save 53.35 (15%)
-
Add to cart
Chemical Identifiers
CAS123334-00-9
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofAFree-Sulfuric Acid: Report Value
Assay from Suppliers CofAWater: Report Value
Assay from Suppliers CofATitanium Oxysulfate: Report value
Titanium(IV) oxide sulfate sulfuric acid hydrate is widely used as a pigment in paints and dyes. It is also useful in the preparation of stable amorphous titanium oxide hollow microspheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only