Thermo Scientific Chemicals

Titanium(IV) bromide, 98%, Thermo Scientific Chemicals

Catalog number: 039592.09
10 g, Each
Thermo Scientific Chemicals

Titanium(IV) bromide, 98%, Thermo Scientific Chemicals

Catalog number: 039592.09
10 g, Each
Quantity
Catalog number: 039592.09
also known as 039592-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-68-6
IUPAC Name
titanium(4+) tetrabromide
Molecular Formula
Br4Ti
InChI Key
UBZYKBZMAMTNKW-UHFFFAOYSA-J
SMILES
[Ti+4].[Br-].[Br-].[Br-].[Br-]
Assay (unspecified)
97.6% min

Description

Titanium(IV) bromide is used as a Lewis acid catalyst in organic synthesis. It reacts with tetrahydrofurnan and 2-methylpyridine and forms adducts such as TiBr4(THF)2 and TiBr4(2-MePy). It is also used for the formation of titanium silicide thin films on silicon substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text