Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis), Thermo Scientific Chemicals
Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2 g
10 g
Catalog number 039630.18
also known as 039630-18
Price (USD)/ Each
1,338.65
Online exclusive
1487.00 
Save 148.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
1,338.65
Online exclusive
1487.00 
Save 148.35 (10%)
Add to cart
Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis), Thermo Scientific Chemicals
Catalog number039630.18
Price (USD)/ Each
1,338.65
Online exclusive
1487.00 
Save 148.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialTantalum(V) tetraethoxide 2,4-pentanedionate
CAS20219-33-4
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
Tantalum(V) tetraethoxide 2,4-pentanedionate is used in pharmaceutical, biochemical and chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only