Thermo Scientific Chemicals

Nickel(II) citrate hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 039634.22
100 g, Each
Thermo Scientific Chemicals

Nickel(II) citrate hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 039634.22
100 g, Each
Quantity
Catalog number: 039634.22
also known as 039634-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6018-92-4
IUPAC Name
trinickel(2+) bis(2-hydroxypropane-1,2,3-tricarboxylate)
Molecular Formula
C12H10Ni3O14
InChI Key
UPPLJLAHMKABPR-UHFFFAOYSA-H
SMILES
[Ni++].[Ni++].[Ni++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Assay from Suppliers CofA
Ni: ≥23.0%
Appearance (Color)
Green
Form
Crystals or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text