Chemical Identifiers
CAS6018-92-4
IUPAC Nametrinickel(2+) bis(2-hydroxypropane-1,2,3-tricarboxylate)
Molecular FormulaC12H10Ni3O14
InChI KeyUPPLJLAHMKABPR-UHFFFAOYSA-H
SMILES[Ni++].[Ni++].[Ni++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more