Zinc peroxide, approximately 50% ZnO{2}, Thermo Scientific Chemicals
Zinc peroxide, approximately 50% ZnO{2}, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc peroxide, approximately 50% ZnO{2}, Thermo Scientific Chemicals

Curing agent, rubber accelerator
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 039635.22
also known as 039635-22
Price (USD)/ Each
35.30
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
35.30
Add to cart
Zinc peroxide, approximately 50% ZnO{2}, Thermo Scientific Chemicals
Catalog number039635.22
Price (USD)/ Each
35.30
-
Add to cart
Chemical Identifiers
CAS1314-22-3
IUPAC Namezinc(2+) dioxidanediide
Molecular FormulaO2Zn
InChI KeyIPTOGCUGCFHDSS-UHFFFAOYSA-N
SMILES[Zn++].[O-][O-]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentAssay ~50%
Zinc peroxide is used as curing agent, rubber accelerator. has been applied in a variety of settings, from medicine to aesthetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only