Sodium stearate, Thermo Scientific Chemicals
Sodium stearate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium stearate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 039648.30
also known as 039648-30
Price (USD)/ Each
43.65
Online exclusive
48.20 
Save 4.55 (9%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
43.65
Online exclusive
48.20 
Save 4.55 (9%)
Add to cart
Sodium stearate, Thermo Scientific Chemicals
Catalog number039648.30
Price (USD)/ Each
43.65
Online exclusive
48.20 
Save 4.55 (9%)
-
Add to cart
Chemical Identifiers
CAS822-16-2
IUPAC Namesodium octadecanoate
Molecular FormulaC18H35NaO2
InChI KeyRYYKJJJTJZKILX-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
CommentFree Fatty Acid (FFA): ≤3.0%
CommentMelting Point: ≥200°C
Loss on Drying≤3.0%
Appearance (Color)White
Used in adhesives and sealants, laundry and dishwashing products, plastic and rubber products. Used as surface active agents. It is the gelling agent for deodrant sticks. Used as waterproofing additives and ointments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only