Thermo Scientific Chemicals

Titanium(III) sulfate, 20% in 1-4% sulfuric acid, Thermo Scientific Chemicals

Catalog number: 039664.30
250 g, Each
Thermo Scientific Chemicals

Titanium(III) sulfate, 20% in 1-4% sulfuric acid, Thermo Scientific Chemicals

Catalog number: 039664.30
250 g, Each
Quantity
Catalog number: 039664.30
also known as 039664-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10343-61-0
IUPAC Name
dititanium(3+) trisulfate
Molecular Formula
O12S3Ti2
InChI Key
SOBXOQKKUVQETK-UHFFFAOYSA-H
SMILES
[Ti+3].[Ti+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Concentration (weight/weight)
19-22%

Description

Titanium(III) sulfate is used in the manufacture of bulk, large scale chemicals (including petroleum products), fine chemicals, basic metals, including alloys, computer, electronic and optical products, electrical equipment. It is also used as a biochemical for proteomics research. It is a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text