Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Type | Reagent |
Standard Concentration | 100% |
Operation | Used for cation exchange eluents for columns packed with sulfonated resins, including the CS3 and CS10. |
Hazards | Irritant |
Standard Components | DL-2,3-Diamino Propionic Acid Mono Hydrochloride |
For Use With (Application) | Cation Exchange Applications |
Quantity | 5 g |
Description | DL-2,3-Diaminopropionic Acid Monohydrochloride (DAP-HCl); 5 g |
Storage | Room Temperature |
Unit Size | Each 1 |
Catalog Number | Specifications | Unit Size | Quantity | For Use With (Application) | Price (USD) | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
039670 | Each 1 | 5 g | Cation Exchange Applications | 534.00 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Dionex DAP-HCl cation reagent contains the following;
Quality to Trust In
Only high-purity, reagent-grade chemicals and water (18MO-cm), quality-control checked for ionic contamination, are used in Thermo Scientific™ Dionex™ reagents, standards and eluent concentrates.