Search Thermo Fisher Scientific
Catalog Number | Quantity | For Use With (Application) |
---|---|---|
039672 | 5 g | Post Column Derivatization of Transition Metals Separated Using the IonPac CS5A Column |
The Dionex 4-(2-Pyridylazo) Resorcinol Monosodium Salt (PAR), 5g is used for post-column derivatization of transition metals separated on the Dionex IonPac CS5A column. This reagent should be used with the Thermo Scientific™ Dionex™ MetPac™ Post Column Diluent.
Share catalog number, name or link