Titanium granules, 99.99% (metals basis excluding Na and K), Thermo Scientific Chemicals
Titanium granules, 99.99% (metals basis excluding Na and K), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium granules, 99.99% (metals basis excluding Na and K), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
500 g
Catalog number 039677.18
also known as 039677-18
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
Add to cart
Titanium granules, 99.99% (metals basis excluding Na and K), Thermo Scientific Chemicals
Catalog number039677.18
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metal99.99% (metals basis excluding Na and K)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only