Boron powder, amorphous, APS <1 micron, 93-96%, Mg 3% max, Thermo Scientific Chemicals
Boron powder, amorphous, APS &lt;1 micron, 93-96%, Mg 3% max, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron powder, amorphous, APS <1 micron, 93-96%, Mg 3% max, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039680.30
Price (USD)/ Each
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7440-42-8
IUPAC Nameboron
Molecular FormulaB
InChI KeyZOXJGFHDIHLPTG-UHFFFAOYSA-N
SMILES[B]
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisBoron, B: ≥92.5 to ≤96.5%
Particle Size≤5.00µm
Appearance (Color)Black to brown
Impurity contentMagnesium, Mg: ≤3.00%
FormPowder

Fullerene is used in organic synthesis, electronics, Sensors, Composites, Energy Storage, Study of Life Science systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only