Thermo Scientific Chemicals

Niobium foil, 0.05mm (0.002in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 039702.HP
100 x 500 mm, Each
Thermo Scientific Chemicals

Niobium foil, 0.05mm (0.002in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 039702.HP
100 x 500 mm, Each
Quantity
Catalog number: 039702.HP
also known as 039702-HP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-03-1
IUPAC Name
niobium
Molecular Formula
Nb
InChI Key
GUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES
[Nb]
Appearance (Color)
Silvery
Assay from Supplier's CofA
≥99.8% (metals basis)
Form
Foil, 0.05mm (0.002in) thick, annealed.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text