Specifications
Chemical Name or MaterialNickel gauze
Name NoteWoven from 0.13mm (0.005 in.) dia. wire
CAS7440-02-0
Health Hazard 1H317-H351-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
View more