Fullerene powder, 97% C{70}, Thermo Scientific Chemicals
Fullerene powder, 97% C{70}, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Fullerene powder, 97% C{70}, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
10 mg
250 mg
Catalog number 039720.MF
also known as 039720-MF
Price (USD)/ Each
-
Quantity:
50 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialFullerene powder
CAS115383-22-7
Health Hazard 1H228-H315-H319-H335
Health Hazard 2GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
Fullerene powder is used in chemical research. It is extensively used for several biomedical applications including the design of high-performance MRI contrast agents, X-Ray imaging contrast agents, photodynamic therapy and drug and gene delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only