Carbon yarn, woven from 0.076mm (0.003in) dia fibers, Thermo Scientific Chemicals
Carbon yarn, woven from 0.076mm (0.003in) dia fibers, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Carbon yarn, woven from 0.076mm (0.003in) dia fibers, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
25 m
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039721.G5
also known as 039721-G5
Price (USD)/ Each
-
Quantity:
5 m
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialCarbon yarn
Name NoteWoven from 0.076mm (0.003 in.) dia. fibers
Recommended StorageAmbient temperatures
EINECS Number231-153-3
TSCAYes
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only