Thermo Scientific Chemicals

Xenon difluoride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 039739.09
10 g, Each
Thermo Scientific Chemicals

Xenon difluoride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 039739.09
10 g, Each
Quantity
Catalog number: 039739.09
also known as 039739-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13709-36-9
IUPAC Name
difluoroxenon
Molecular Formula
F2Xe
InChI Key
IGELFKKMDLGCJO-UHFFFAOYSA-N
SMILES
F[Xe]F
Total Metal Impurities
0.5% max.

Description

Used as Fluorinating agent and oxidizing agent, gaseous etchant for silicon, particularly in the production of microelectromechanical systems (MEMS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text