Thermo Scientific Chemicals

Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 039741.06
5 g, Each
Thermo Scientific Chemicals

Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 039741.06
5 g, Each
Quantity
Catalog number: 039741.06
also known as 039741-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15283-45-1
IUPAC Name
gold(1+) trisodium disulfanidesulfonate
Molecular Formula
AuNa3O6S4
InChI Key
FJCMCHNDCIYVST-UHFFFAOYSA-J
SMILES
[Na+].[Na+].[Na+].[Au+].[O-]S([S-])(=O)=O.[O-]S([S-])(=O)=O
Appearance (Color)
White
Assay (special)
Au: ≥34.8%
Form
Powder
Total Metal Impurities
≤0.1%

Description

Gold(I) sodium thiosulfate hydrate is useful in organogold compounds. Further, it serves as an antirheumatic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text