Gold(III) acetate, 99.9% (metals basis), Thermo Scientific Chemicals
Gold(III) acetate, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold(III) acetate, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
1 g
5 g
Catalog number 039742.02
also known as 039742-02
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
0.5 g
Request bulk or custom format
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
Add to cart
Gold(III) acetate, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number039742.02
Price (USD)/ Each
138.65
Online exclusive
154.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS15804-32-7
IUPAC Namegold(3+) triacetate
Molecular FormulaC6H9AuO6
InChI KeyOTCKNHQTLOBDDD-UHFFFAOYSA-K
SMILES[Au+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
FormPowder
Total Metal Impurities≤0.1%
Used in solar energy technologies. Used in petrochemical cracking and automotive catalysts, water treatment, plating, textiles, research and in optic, laser, crystal and glass applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only