Thermo Scientific Chemicals

Gold(III) acetate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 039742.03
1 g, Each
Thermo Scientific Chemicals

Gold(III) acetate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 039742.03
1 g, Each
Quantity
Catalog number: 039742.03
also known as 039742-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15804-32-7
IUPAC Name
gold(3+) triacetate
Molecular Formula
C6H9AuO6
InChI Key
OTCKNHQTLOBDDD-UHFFFAOYSA-K
SMILES
[Au+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Appearance (Color)
Brown
Form
Powder
Total Metal Impurities
≤0.1%

Description

Used in solar energy technologies. Used in petrochemical cracking and automotive catalysts, water treatment, plating, textiles, research and in optic, laser, crystal and glass applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text