Lead zirconium titanium oxide, polymeric precursor, oxide ≈30 wt%, Thermo Scientific Chemicals
Lead zirconium titanium oxide, polymeric precursor, oxide ≈30 wt%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead zirconium titanium oxide, polymeric precursor, oxide ≈30 wt%, Thermo Scientific Chemicals

Used in piezoelectric devices and electro-optic devices
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 039758.09
also known as 039758-09
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS12626-81-2
IUPAC Nametitanium(4+) zirconium(4+) λ²-lead(2+) pentaoxidandiide
Molecular FormulaO5PbTiZr
InChI KeyHFGPZNIAWCZYJU-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[O--].[Ti+4].[Zr+4].[Pb++]
View more
SpecificationsSpecification SheetSpecification Sheet
Formviscous liquid to semi - solid
Assay (unspecified)Oxide > 25 wt%
Appearance (Color)Pale yellow to brown
Lead zirconium titanium oxide is used in piezoelectric devices and electro-optic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only