Ethylenediaminetetraacetic acid disodium salt, 0.100N (0.050M) Standardized solution, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid disodium salt, 0.100N (0.050M) Standardized solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid disodium salt, 0.100N (0.050M) Standardized solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number 039781.K2
also known as 039781-K2
Price (USD)/ Each
42.65
Online exclusive
46.90 
Save 4.25 (9%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
42.65
Online exclusive
46.90 
Save 4.25 (9%)
Add to cart
Ethylenediaminetetraacetic acid disodium salt, 0.100N (0.050M) Standardized solution, Thermo Scientific Chemicals
Catalog number039781.K2
Price (USD)/ Each
42.65
Online exclusive
46.90 
Save 4.25 (9%)
-
Add to cart
Chemical Identifiers
CAS139-33-3
IUPAC Namedisodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Molecular FormulaC10H14N2Na2O8
InChI KeyZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofAEDTA Disodium: 0.1N = 0.05M
FormLiquid
Assay from Supplier's CofA0.1000N ? 0.0002N
Appearance (Color)Colorless
It is widely used in textile industry. Usually applied to dissolve limescale. It is applied in textile industry, pulp and paper industry and also in chelation therapy. In cosmetics it acts as a sequestering agent. It acts as a corrosion inhibitor to carbon steel in the industries. It also acts as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only