KBr Quick Press Kit with 7mm die set
KBr Quick Press Kit with 7mm die set
Thermo Scientific Chemicals

KBr Quick Press Kit with 7mm die set

 
Have Questions?
Catalog number 039786.KT
also known as 039786-KT
Price (USD)/ Each
1,407.65
Online exclusive
1564.00 
Save 156.35 (10%)
-
Add to cart
Request bulk or custom format
Price (USD)/ Each
1,407.65
Online exclusive
1564.00 
Save 156.35 (10%)
Add to cart
KBr Quick Press Kit with 7mm die set
Catalog number039786.KT
Price (USD)/ Each
1,407.65
Online exclusive
1564.00 
Save 156.35 (10%)
-
Add to cart
Specifications
TypePress Kit
Includes7mm die set
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only