Thermo Scientific Chemicals

Potassium bromide, FTIR Grade, Thermo Scientific Chemicals

Catalog number: 039794.14
25 g, Each
Thermo Scientific Chemicals

Potassium bromide, FTIR Grade, Thermo Scientific Chemicals

Catalog number: 039794.14
25 g, Each
Quantity
Catalog number: 039794.14
also known as 039794-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-02-3
IUPAC Name
potassium bromide
Molecular Formula
BrK
InChI Key
IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES
[K+].[Br-]
Comment
Potassium bromide, FTIR Grade powder is suitable for IR and FTIR spectroscopy

Description

Potassium bromide is used as a veterinary drug, as an antiepileptic medication for dogs. It used for removal of peripheral membrane proteins. Potassium bromide and sodium perborate play an important role for mild and regioselective oxidative bromination of anilines. Its Pellet is useful for fine structural characteristics by infared spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text